Kimberly-Clark Professional™ KleenGuard™ A40 Liquid and Particle Protection Shoe Covers
A40 shoe covers are made with breathable, microporous film to protect from debris, non-hazardous liquid chemicals, and dry particulates.
Supplier:  Kimberly-Clark Professional™ 44490
 View more versions of this product
Catalog No. 19-120-2410
Explore available promotions
Description
Description
  • Made of microporous film laminate
  • Serged seams
  • Choose from fully elasticized formed fit or elastic opening styles
  • Ambidextrous single use cover 5.25 in. or 7 in. height
  • Made to keep out debris, dry particulates, and some liquids
  • Adheres to NFPA 99 criteria for antistatic materials
Compliance
Meets NFPA™ 99 Health Care Facilities criteria for antistatic materials.
Specifications
Specifications
Universal
Microporous Film Laminate
Single-use
Fully Elastic
Serged
NFPA 99
7 in.
Shoe Covers
Microporous Fabric
White
Non-sterile
44490
Manufacturing, Painting, Cleaning, Liquid Handling, Utilities, Maintenance
400/Cs.
Fluid