missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ A40 Liquid and Particle Protection Shoe Covers
A40 shoe covers are made with breathable, microporous film to protect from debris, non-hazardous liquid chemicals, and dry particulates.
Supplier: Kimberly-Clark Professional™ 44494
Description
- Made of microporous film laminate
- Serged seams
- Choose from fully elasticized formed fit or elastic opening styles
- Ambidextrous single use cover 5.25 in. or 7 in. height
- Made to keep out debris, dry particulates, and some liquids
- Adheres to NFPA 99 criteria for antistatic materials
Compliance
Meets NFPA™ 99 Health Care Facilities criteria for antistatic materials.
Specifications
X-Large | |
Microporous Film Laminate | |
Single-use | |
Elastic Top | |
Serged | |
NFPA 99 | |
5.25 in. | |
Shoe Covers |
Microporous Fabric | |
White | |
Non-sterile | |
44494 | |
Manufacturing, Painting, Cleaning, Liquid Handling, Utilities, Maintenance | |
400/Cs. | |
Fluid |