missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ A40 Liquid and Particle Protection Coveralls
Protective coveralls are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
Supplier: Kimberly-Clark Professional™ 44326
Description
Protective coveralls are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
- Breathable, microporous film laminate fabric.
- Barrier protection against hazardous particulates and aerosols and against low-hazard liquid splash.
- ASTM F1670 for blood penetration and ASTM F1671 for bloodborne pathogens.
- ASTM F903 Penetration Testing provides passing results for isocyanate paint (40%) and Potassium Chromate (20%).
- Anti-stat according to NFPA 99.
- Features a zippered front and come in a variety of sizes.
- Eligible for recycling via RightCycle by Kimberly-Clark Professional™ when used in non-hazardous applications as classified by federal, state, and local regulations.
- Various configurations available, including open or elastic wrists and ankles, elastic back, a hood, and non-skid boots.
- Human-centric Reflex™ design offers comfortable fit and freedom of movement.
Compliance
Meets ANSI/ISEA 101-1996; ASTM F1670 testing for penetration of blood and bodily fluids; NFPA 99 healthcare facilities criteria for antistatic materials.
Specifications
General Purpose Coveralls | |
Unisex | |
White | |
Elastic | |
Hooded | |
Front Zipper | |
Particle, Liquid | |
KleenGuard™ A40 | |
Elastic Wrists, Ankles with Hood |
3X-Large | |
Microporous Film Laminate | |
Limited-use | |
Elastic | |
Elastic | |
Non-sterile | |
ASTM F1670, ASTM F1671, ASTM F903, NFPA 99 | |
Fiberglass Manufacturing, Liquid Handling, Utilities, Pressure Washing, Tank Cleaning, Maintenance and General Manufacturing |