Kimberly-Clark Professional™ KleenGuard™ A40 Liquid and Particle Protection Coveralls
Protective coveralls are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
Supplier:  Kimberly-Clark Professional™ 44337
 View more versions of this product
Catalog No. 19-120-2401G
Explore available promotions
Description
Description
Protective coveralls are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
  • Breathable, microporous film laminate fabric.
  • Barrier protection against hazardous particulates and aerosols and against low-hazard liquid splash.
  • ASTM F1670 for blood penetration and ASTM F1671 for bloodborne pathogens.
  • ASTM F903 Penetration Testing provides passing results for isocyanate paint (40%) and Potassium Chromate (20%).
  • Anti-stat according to NFPA 99.
  • Features a zippered front and come in a variety of sizes.
  • Eligible for recycling via RightCycle by Kimberly-Clark Professional™ when used in non-hazardous applications as classified by federal, state, and local regulations.
  • Various configurations available, including open or elastic wrists and ankles, elastic back, a hood, and non-skid boots.
  • Human-centric Reflex™ design offers comfortable fit and freedom of movement.
Compliance
Meets ANSI/ISEA 101-1996; ASTM F1670 testing for penetration of blood and bodily fluids; NFPA 99 healthcare facilities criteria for antistatic materials.
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Hooded
Front Zipper
Particle, Liquid
KleenGuard™ A40
Elastic Wrists, Ankles with Hood and Boots
4X-Large
Microporous Film Laminate
Limited-use
Elastic
Elastic
Non-sterile
ASTM F1670, ASTM F1671, ASTM F903, NFPA 99
Fiberglass Manufacturing, Liquid Handling, Utilities, Pressure Washing, Tank Cleaning, Maintenance and General Manufacturing