Kimberly-Clark Professional™ KleenGuard™ A70 Chemical Spray Protection Smock
Polyethylene-coated fabric
Supplier: Kimberly-Clark Professional™ 09829
Description
Resistant to many water-based liquids, greases and oils, and liquid chemical sprays
- Elastic wrists
- Tie-front closure at waist
- Back opening
- Yellow
Specifications
Polyethylene | |
Yellow | |
44 in. |