Kimberly-Clark Professional™ Smith & Wesson™ Magnum 3G™ Eyewear
Ergonomically dimensioned three-point fit for comfort
Supplier:  Kimberly-Clark Professional™ 19961
 View more versions of this product
Catalog No. 19-150-2000
Explore available promotions
Description
Description
Ergonomically dimensioned three-point fit for comfort
  • Built-in hinge stops help to eliminate scratches
  • Self-adjusting ergonomic rubber nose pad with rubber fingers
  • Mini-Magnum™ designed to fit small faces
  • Provides 99.9% UV protection
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Specifications
Oval
Clear
Platinum
Platinum (Frame)
Meets ANSI Z87.1-2003 standards.
Safety Glasses
Full-sized