missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark™ WypAll™ X90 Cloth Wipers

Reusable cloths that are ideal for tough maintenance and cleaning tasks. Kimberly-Clark™ WypAll™ X90 Cloth Wipers are strong, durable, highly absorbent and solvent resistant.

Supplier:  Kimberly-Clark™ 12889

 View more versions of this product

Catalog No. 18-892-193


Explore available promotions

Description

Description

  • Releases 90% of the solvent absorbed, reducing your solvent spend
  • Reduces product waste by up to 35%, helping you to manage spend and wastage
  • Advanced Hydroknit™ technology ensures each wipe is strong, durable, highly absorbent and solvent resistant
  • Portable and ideal for small work spaces
  • Designed for tough manufacturing processes where low lint and high absorbency are critical
  • Designed for tough maintenance and cleaning tasks in industrial manufacturing
  • Designed to help you improve productivity and efficiency whilst reducing waste
Specifications

Specifications

Heavy-duty Wiper
34 cm
28.19 cm
Blue
Ideal for heavy wiping, prepping surfaces with solvents and cleaning Metal shaving
13.4 in.
11.1 in.
Non-woven Polyproylene Cellulose
13.4 x 11.1 in. (34 x 28.19 cm)
SDS
Documents

Documents

Promotions

Promotions