Learn More
DWK Life Sciences Kimble™ KIMAX™ Accuform™ Micro-Vial™ Vials
Conical cavity allows maximum retrieval of contents with a syringe needle.
Supplier:  DWK Life Sciences 607102
 View more versions of this product
Catalog No. 06-100J
Only null left
Explore available promotions
Description
Description
  • Clear glass Micro-Vial with open top closure and PTFE-faced silicone septum
  • Made with extra thick clear glass walls for both sturdiness and magnification of contents
  • Ungraduated, with marking spot
  • Tube has a screw thread finish
Specifications
Specifications
PTFE/Silicone
21 mm
2.0 mL
Tapered
0.5 dr.
40 mm
Clear
Borosilicate Glass
No
Screw
Screw Cap (Open-top)
ASTM E438, USP Type I, Type I, Class A
Product Suggestions
Product Suggestions