DWK Life Sciences Kimble™ KIMAX™ Caps for Media Bottles
Black phenolic or propylene caps
Supplier:  DWK Life Sciences 7380238430
 View more versions of this product
Catalog No. 06-422-1B
Explore available promotions
Description
Description
  • Phenolic caps with plain 14B rubber liners or PTFE-faced 14B rubber liners
  • Polypropylene caps with bonded PTFE-faced silicone liners to eliminate glue contamination and loose liners
Specifications
Specifications
Black
144 Cs.
Media bottle
Phenolic Resin
38-430
PTFE/14B Rubber