DWK Life Sciences Kimble™ KIMAX™ Caps for Media Bottles
Black phenolic or propylene caps
Supplier: DWK Life Sciences 7380333430
Description
- Phenolic caps with plain 14B rubber liners or PTFE-faced 14B rubber liners
- Polypropylene caps with bonded PTFE-faced silicone liners to eliminate glue contamination and loose liners
Specifications
Black | |
144 Cs. | |
Media bottle |
Phenolic Resin | |
33-430 | |
14B Rubber |