DWK Life Sciences Kimble™ KIMAX™ Cell Culture/Media Erlenmeyer Flasks with GL45 Screw Thread
Large opening
Supplier: DWK Life Sciences 267201000
Description
Large opening
- Blue PP caps
- Clear seal rings
- Low extractable borosilicate glass
Specifications
1000 mL | |
Borosilicate Glass | |
250-1000 | |
205 mm |