DWK Life Sciences Kimble™ KIMAX™ Distilling Apparatus Flat-Bottom Flask
Designed for general purpose distillation and phenol distillation
Supplier:  DWK Life Sciences 21500G500
 View more versions of this product
Catalog No. 13-631-8
Explore available promotions
Description
Description
Specifications
Specifications
Designed for general purpose distillation and phenol distillation
16.9 oz., 500mL
22
16.9 oz.
Distillation Apparatus Flask
500mL
19/38