DWK Life Sciences Kimble™ KIMAX™ Filling Funnels
Fire-polished edges reduce chipping
Supplier:  DWK Life Sciences 2902080
 View more versions of this product
Catalog No. 10-346-5B
Explore available promotions
Description
Description
Fire-polished edges reduce chipping
  • Material: 33 expansion borosilicate glass
  • Fire-polished edges
  • Clear
Specifications
Specifications
8cm
Borosilicate Glass
3.5cm
1.5cm
ASTM E1095, Type I, Class A