DWK Life Sciences Kimble™ KIMAX™ Filtering Flasks with Side Arm
Heavy walled with graduated scale, side tubulation, tooled neck
$335.88 - $1380.47
Specifications
Certifications/Compliance | ASTM E1406, Type II, Class I |
---|---|
Material | Borosilicate Glass |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
10-181G
|
DWK Life Sciences
270602000 |
2000 mL |
Case for $335.88
|
|
|||||
10-181A
|
DWK Life Sciences
2706025 |
25 mL |
Case of 18 Each for $1,380.47
|
|
|||||
10-181B
|
DWK Life Sciences
2706050 |
50 mL |
Pack of 6 Each for $562.32
Case of 3 Pack for $1,686.96
|
|
|||||
10-181D
|
DWK Life Sciences
27060250 |
250 mL |
Pack of 6 Each for $377.59
Case of 3 Pack for $1,132.77
|
|
|||||
10-181F
|
DWK Life Sciences
270601000 |
1000 mL |
Pack of 6 Each for $1,014.88
Case of 2 Pack for $2,029.76
|
|
|||||
10-181E
|
DWK Life Sciences
27060500 |
500 mL |
Pack of 6 Each for $613.59
Case of 3 Pack for $1,840.77
|
|
|||||
10-181H
|
DWK Life Sciences
270604000 |
4 L |
Case for $587.00
|
|
|||||
Description
Heavy walled with graduated scale, side tubulation, tooled neck
- Sizes 25, 50, and 125mL suitable for microanalysis
- Sizes 25 and 500mL have side hose connections for 0.3125 in. I.D. (8mm) flexible tubing
- Sizes 1000mL and larger accept 0.375 in. I.D. (9.5mm) tubing
Compliance
Designed from ASTM™ E 1406, Type II requirements
Specifications
ASTM E1406, Type II, Class I | |
Borosilicate Glass |