DWK Life Sciences Kimble™ KIMAX™ Flasks Serialized and Class A Certified with Snap Caps
Calibrated and certified against NIST-Traceable equipment
Supplier:  DWK Life Sciences 28012100
 View more versions of this product
Catalog No. 10-201B
Explore available promotions
Description
Description
Calibrated and certified against NIST-Traceable equipment
  • Permanent individual serial no.
  • Designed according to ASTM E 288
  • To contain
Specifications
Specifications
Polyethylene
Borosilicate Glass
16 cm
4
100 mL
6.2 in.
±0.08 mL