DWK Life Sciences Kimble™ KIMAX™ Organic Chemistry Kits
Include an assortment of items required for basic organic laboratory experiments
Supplier:  DWK Life Sciences 2698000000
 View more versions of this product
Catalog No. K269800-0000
Explore available promotions
Description
Description
  • For education and research
  • All joints in a kit are the same standard taper size, making it easy to set up a number of procedures
Order Info
Kit 11-301-20 comes in a heavyweight corrugated cardboard and inserts are soft foam. Kit components are listed in the chart below; contact your Fisher Scientific Customer Service Representative to order replacement items.
Specifications
Specifications
Standard Taper Kit
For extraction, reflux, reflux with addition, fractionation, and distillation
19/22
USP Type I and ASTM E438 Type I, Class A
Bleed adapter, claisen adapter, condenser, connecting adapter, distillation column, 25mL, 50mL, 100mL, 250mL, 500mL Flask, 125mL PTFE funnel, outlet adapter, stopper, vacuum adapter