DWK Life Sciences Kimble™ KIMAX™ Soxhlet Apparatus with Allihn Condenser, Condenser only
For use with for Kimble™ extraction apparatus
Supplier:  DWK Life Sciences 2402740
 View more versions of this product
Catalog No. 09-551-10B
Explore available promotions
Description
Description
  • The soxhlet extractor was originally designed for the extraction of a lipid from a solid material
  • Not limited to the extraction of lipids - typically, a soxhlet extraction is only required where the desired compound has a limited solubility in a solvent and the impurity is insoluble in that solvent
  • If the desired compound has a high solubility in a solvent then a simple filtration can be used to separate the compound from the insoluble substance
Order Info
See the Complete Assembly (09-551A, 09-551B, 09-551C) which contains a condenser, extraction tube and flask.
Specifications
Specifications
Extractor Condenser
45/50
4 cm
Soxhlet Extraction Apparatus
1.57 in.