missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ KIMAX™ Squibb Pear-Shaped Sep. Funnel with PTFE-Plug Stopcock, standard taper PE Stopper

Closed-bottom, linear high-density polyethylene stopper in standard taper ground neck

Supplier:  DWK Life Sciences 29049F250

 View more versions of this product

Catalog No. 10-437-11D



Explore available promotions

Description

Description

Closed-bottom, linear high-density polyethylene stopper in standard taper ground neck

Lower stem made of large-diameter tubing for liquid to drain out completely when stopcock is closed.

Order Info

Stem lengths, 60mm.

Compliance

Designed from ASTM™ E 1096-86.

Specifications

Specifications

250 mL
ASTM E1096, Type IV
2 Pack
HDPE
Squibb Separatory Funnel
Borosilicate Glass
60mm
4; PTFE
22
Videos
SDS
Documents

Documents

Promotions

Promotions