missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ KIMAX™ Storage/Media Bottles

With linerless blue polypropylene caps

Supplier:  DWK Life Sciences KC14395100

 View more versions of this product

Catalog No. 12-141-320


Explore available promotions

Description

Description

With linerless blue polypropylene caps

  • KimCote™ plastic coating helps prevent and contain spills in the event of glassware breakage
  • GL 45 screw thread bottles with white graduations and marking spots
  • Wide opening for easy access (approx. 30mm I.D.)
  • Linerless cap is blue polypropylene and autoclavable (140°C max)
  • Replaceable pour ring included
  • Ultra-clear for improved sample viewing
  • Non-slip surface — wet or dry

Applications:

Pressurized glass, chemical storage, transportation of samples, handling of hazardous chemicals, harmful biologicals and acids

Specifications

Specifications

3.3 oz., 100 mL
3.3 oz.
2.2 in.
30 mm
Plastic
20 to 80mL
100 mm
Polypropylene
56 mm
100 mL
Ultra Clear
1.18 in.
Clear drip-free Polypropylene pouring ring
Blue
Yes
3.9 in.
GL 45
Screw Cap
Videos
SDS
Documents

Documents

Promotions

Promotions