missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ KIMAX™ Storage/Media Bottles
With linerless blue polypropylene caps
Supplier: DWK Life Sciences KC143951000
Description
With linerless blue polypropylene caps
- KimCote™ plastic coating helps prevent and contain spills in the event of glassware breakage
- GL 45 screw thread bottles with white graduations and marking spots
- Wide opening for easy access (approx. 30mm I.D.)
- Linerless cap is blue polypropylene and autoclavable (140°C max)
- Replaceable pour ring included
- Ultra-clear for improved sample viewing
- Non-slip surface — wet or dry
Applications:
Pressurized glass, chemical storage, transportation of samples, handling of hazardous chemicals, harmful biologicals and acids
Specifications
| 33.8 oz., 1000 mL | |
| 33.8 oz. | |
| 3.9 in. | |
| 30 mm | |
| Plastic | |
| 100 to 900mL | |
| 225 mm | |
| Polypropylene | |
| 101 mm |
| 1000 mL | |
| Ultra Clear | |
| 1.18 in. | |
| Clear drip-free Polypropylene pouring ring | |
| Blue | |
| Yes | |
| 8.8 in. | |
| GL 45 | |
| Screw Cap |