DWK Life Sciences Kimble™ KIMAX™ Titration Flasks
Wide mouth
Supplier:  DWK Life Sciences 26650125
 View more versions of this product
Catalog No. 10-091A
Explore available promotions
Description
Description
Wide mouth
  • Heavy-duty tooled top finish
  • Thick walls
  • Long tapered outside contour
  • Durable white graduations on side
  • Stoppers not included
Compliance
Designed from ASTM™ E 1404, Type I, Class 2 requirements
Specifications
Specifications
Top style: Heavy duty tooled with capacity scale
125 mL
50 to 125 mL
4.25 in.
Wide
6
2.59 in.
Borosilicate Glass
25 mL
10.8 cm
66 mm
SDS
Documents
Documents
Accessories