DWK Life Sciences Kimble™ Kontes™ 24/40 Center Neck Cyanide Distillation Apparatus
Center neck cyanide distillation apparatus 24/40. This apparatus is used in the analysis of industrial waste waters, saline and reagent grade water.
Supplier:  DWK Life Sciences 4791000000
Catalog No. K479100-0000
Explore available promotions
Description
Description
  • Cyanide as hydrocyanic acid (HCN) is released from cyanide complexes by means of reflux-distillation and absorbed in a scrubber containing a sodium hydroxide solution
  • The cyanide ion in the absorbing solution is then determined colorimetrically or by volumetric titration
  • Heating mantle (pictured) is not included
  • Capacity 1000mL
  • Overall Height 500mm, Overall Width 450mm
  • Feature 45/50 Upper Standard Taper
  • Fits Tubing ID 3/8 in.
  • Case of 1
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
USP Type I, ASTM E438, Type I, Class A
Quantitative analysis of various industrial wastewater, saline and reagent grade water
19.68 in.
1000 mL
33.81 oz.
50 cm