DWK Life Sciences Kimble™ Kontes™ 19/38 Cyanide Distillation Apparatus
Apparatus for quantitative analysis of various industrial waste waters, saline and reagent grade water.
Supplier:  DWK Life Sciences 4791100000
Catalog No. K479110-0000
Explore available promotions
Description
Description
  • Cyanide as hydrocyanic acid (HCN) is released from cyanide complexes by means of reflux-distillation and absorbed in a scrubber containing a sodium hydroxide solution
  • The cyanide ion in the absorbing solution is then determined colorimetrically or by volumetric titration
  • Design features compact Standard Taper 19/38 joints
  • Capacity 1000mL
  • Feature Upper Standard Taper
  • Standard Taper joints 19/38
  • Case of 1
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
USP Type I, ASTM E438, Type I, Class A
Quantitative analysis of various industrial wastewater, saline and reagent grade water