missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ Kontes™ Double Vacuum/Gas Manifold

Grease-free operation

$1839.29 - $5369.97

Specifications

Material Borosilicate Glass
View More Specs

Products 5
Catalog Number Mfr. No. Type For Use With (Application) Includes No. of Ports Price Quantity  
Catalog Number Mfr. No. Type For Use With (Application) Includes No. of Ports Price Quantity  
K216050-0000
View Documents Promotion Details
DWK Life Sciences
2160500000
Vacuum Gas Manifold Connecting inert gas and vacuum sources to sidearm stopcocks Metering valves, Pressure/vacuum retainers 3
Each for $1,839.29
 
K216050-0005
View Documents Promotion Details
DWK Life Sciences
2160500005
Vacuum Gas Manifold Connecting inert gas and vacuum sources to sidearm stopcocks Metering valves, Pressure/vacuum retainers 5
Each for $2,871.56
 
K216060-0003
View Documents
DWK Life Sciences
2160600003
Vacuum Gas Manifold Connecting inert gas and vacuum sources 4mm PTFE vacuum valves 3
Each for $2,521.15
 
K216060-0000
View Documents Promotion Details
DWK Life Sciences
2160600000
Vacuum Gas Manifold Connecting inert gas and vacuum sources 4mm PTFE vacuum valves 5
Each for $3,409.68
 
K216040-0004
View Documents Promotion Details
DWK Life Sciences
2160400004
Double Vacuum Gas Manifold Grease-free operation HI-VAC forward-facing valves 4
Each for $5,369.97
 
Description

Description

K216040 Series Vacuum/Gas Manifold

  • Forward-facing valves
  • Compact for assembly in fume hood
  • Modular and easily disassembled for cleaning
  • Ports spaced 3.93 in. (100mm) apart
K215050 Series Vacuum/Gas Manifold
  • An all-glass manifold for connecting inert gas and vacuum sources to sidearm stopcocks
  • Each stopcock has a metering groove cut into the plug to provide fine control of vacuum and inert gases
  • Distance between stopcocks is 2.55 in. (65mm) for the 3 position size and 3.93 in. (100mm) for the 5 position size
  • Supplied with proprietary pressure/vacuum retainers
K216060 Series Vacuum/Gas Manifold
  • For connecting inert gas and vacuum sources
  • Sample contacts only PTFE and borosilicate glass
  • Beams 0.5 in. (12.7mm) O.D. with upper-left and lower-right ends open
  • Delivery ports 9mm O.D.
  • Distance between valves 0.35 in. (100mm)

Specifications

Specifications

Borosilicate Glass
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'