DWK Life Sciences Kimble™ Kontes™ FlexColumn™ Economy Columns
Ideal for use with FPLC™ systems, peristaltic pumps, or gravity flow to 15psi
$92.02 - $706.23
Specifications
Diameter (Metric) Inner 15 mm
Type Economy Chromatography Column
Endcapped Yes
For Use With (Application) Ion exchange, affinity, gel filtration and hydrophobic interaction media
Temperature (Metric) Range 0 to 50°C
View More Specs
Products 17
Catalog Number Mfr. No. Length (Metric) Column Capacity Quantity Price Quantity  
Catalog Number Mfr. No. Length (Metric) Column Capacity Quantity Price Quantity  
K420400-1505
View Documents Promotion Details
DWK Life Sciences
4204001505
50 mm 9 mL 5/Cs.
Pack of 5 Each for $289.64
K420400-1510
View Documents Promotion Details
DWK Life Sciences
4204001510
100 mm 18 mL 5/Cs.
Pack of 5 Each for $348.98
K420400-1512
View Documents
DWK Life Sciences
4204001512
1200 mm 230 mL 3/Cs.
Case of 3 Each for $706.23
K420400-1520
View Documents Promotion Details
DWK Life Sciences
4204001520
200 mm 35 mL 5/Cs.
Pack of 5 Each for $349.98
K420400-1530
View Documents Promotion Details
DWK Life Sciences
4204001530
300 mm 53 mL 5/Cs.
Pack of 5 Each for $349.98
K420400-1515
View Documents Promotion Details
DWK Life Sciences
4204001515
150 mm 27 mL 5/Cs.
Pack of 5 Each for $348.98
K420400-1575
View Documents Promotion Details
DWK Life Sciences
4204001575
750 mm 124 mL 3/Cs.
Pack of 3 Each for $529.75
K420400-1550
View Documents Promotion Details
DWK Life Sciences
4204001550
500 mm 89 mL 5/Cs.
Pack of 5 Each for $464.70
K420401-1505
View Documents Promotion Details
DWK Life Sciences
4204011505
50 mm 9 mL Each
Each for $92.02
K420401-1510
View Documents Promotion Details
DWK Life Sciences
4204011510
100 mm 18 mL Each
Each for $106.09
K420401-1511
View Documents Promotion Details
DWK Life Sciences
4204011511
1000 mm 177 mL Each
Each for $279.19
K420401-1515
View Documents Promotion Details
DWK Life Sciences
4204011515
150 mm 27 mL Each
Each for $106.09
K420401-1520
View Documents Promotion Details
DWK Life Sciences
4204011520
200 mm 35 mL Each
Each for $123.60
K420401-1550
View Documents Promotion Details
DWK Life Sciences
4204011550
500 mm 89 mL Each
Each for $151.47
K420401-1530
View Documents Promotion Details
DWK Life Sciences
4204011530
300 mm 53 mL Each
Each for $121.68
K420401-1575
View Documents Promotion Details
DWK Life Sciences
4204011575
750 mm 124 mL Each
Each for $251.79
K420401-1517
View Documents Promotion Details
DWK Life Sciences
4204011517
1700 mm 301 mL Each
Each for $414.89
Description
Description
Ideal for use with FPLC™ systems, peristaltic pumps, or gravity flow to 15psi
Use columns with ion-exchange, affinity, gel filtration, and hydrophobic interaction media.
  • Durable borosilicate glass
  • With flow adapters
  • Precision-ground column ends form positive seals with endcaps so they don't leak or “creep” under pressure
  • Threaded, easy to remove reservoir cap won't pop off under pressure
  • Reservoir accepts accessory funnels and glass reservoirs
  • Luer-Lock polypropylene inlet and outlet fittings
  • Bed support has a 20μm polyethylene disc that retains most media for low-pressure aqueous chromatography
  • Columns accept flow adapters for adjusting Flex-Column bed height (order adapters separately)
Specifications
Specifications
15 mm
Yes
0 to 50°C
20 μm
HDPE
1.77cm3
Economy Chromatography Column
Ion exchange, affinity, gel filtration and hydrophobic interaction media
14.5 psi
Borosilicate Glass Type I Class A
Male luer lock
2N NaOH or 100% Ethanol
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'