DWK Life Sciences Kimble™ Kontes™ Chromatography Column ETFE Ferrule for 1/8 in. Tubing
Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX™, FLEX-COLUMN™, DISPOSAFLEX™ columns, and ULTRA-WARE™ filtration assemblies.
Supplier: DWK Life Sciences 4208220018
Description
- EFTE Flangeless Ferrule for 1/8 in. OD Tubing
- For use with 420821 Series Male Nuts
- Case of 10
- Replacement Parts in ULTRA-WARE™ Standard Fittings Kit
Specifications
HPLC cap system | |
PTFE | |
Ferrule | |
1 Cs. |