missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Kimble™ Kontes™ HPLC Solvent Pickup Ultra-Ware™ Adapters: Solvent Bottle Adapter
For use with five valve, four valve and filtration caps
Supplier: DWK Life Sciences 9539070000
Description
- GL-45 solvent bottle adapter is made from PBT and converts Ultra-ware GL-45 mobile phase caps to standard 4L solvent bottles for direct access to HPLC instruments
Specifications
Solvent Bottle Adapter | |
PBT |