DWK Life Sciences Kimble™ Kontes™ Micro-Vial Graduated Screw Thread Vials
With solid-top closure and PTFE-faced white rubber liner
Supplier:  DWK Life Sciences 607021
 View more versions of this product
Catalog No. 06-100-1J
Explore available promotions
Description
Description
  • Accuform™
  • KG-33 borosilicate glass
  • Extra thick clear glass walls
Specifications
Specifications
PTFE/Rubber
14 mm
1 mL
1.92 in.
Screw Cap (Open-Top)
12 Cs.
Clear
Borosilicate Glass
Yes
Tapered
49 mm