DWK Life Sciences Kimble™ Kontes™ Microflex™ Microscale Kits: Cold Finger Condenser
Supplier:  DWK Life Sciences 7460650000
Catalog No. 22-300624
Explore available promotions
Description
Description
Condenser and Inlet/Vacuum Adapter (K746035-0814). With standard taper 14/10 joint and 3 connections for 5/16 in. I.D. (7.9mm) hose.
Specifications
Specifications
14/10
0.6 cm
1 Case
Used as an accessory for microscale standard taper 14/10 threaded kits
0.25 in.
Cold Finger Condenser
Borosilicate Glass