DWK Life Sciences Kimble™ Kontes™ Microflex™ Microscale Kits: Hickman Distillation Head
For less than 1 to 3mL.
Supplier:  DWK Life Sciences 7460700000
Catalog No. K746070-0000
Explore available promotions
Description
Description
  • With GPI 20-400 thread and standard taper 14/10 joint.
Specifications
Specifications
Hickman Distillation Head
Borosilicate Glass
14/10
USP Type I and ASTM E438 Type I, Class A
20-400