DWK Life Sciences Kimble™ Kontes™ MIDI-VAP™ 4000 Cyanide Glassware
Replacement cyanide glassware set for the Kimble MIDI-VAP 4000 Cyanide Distillation System (sold separately).
Supplier: DWK Life Sciences 4794600000
Description
- Includes cold finger condenser, distilling head, reaction/absorption tube, absorption impinger
- Set of 1
Specifications
Cold finger condenser, distilling head, reaction/absorption tube, absorption impinger | |
Set of 1 | |
MIDI-VAP™ 4000 distillation systems |
Cyanide Glassware Set | |
Glass | |
U.S. EPA |