DWK Life Sciences Kimble™ Kontes™ Straight with Tip-O-Ring Glass Plugs
Uses glass plug for applications which do not require out-gassing plug material
$100.41 - $161.29
Specifications
Material Glass
Type Replacement Plug
View More Specs
Alert:
Polyacetal knob is not recommended for use over 110°C.
Products 2
Catalog Number Mfr. No. Plug Stopcock No Price Quantity  
Catalog Number Mfr. No. Plug Stopcock No Price Quantity  
K826451-0008
View Documents Promotion Details
DWK Life Sciences
8264510008
8
Each for $100.41
K826451-0004
View Documents Promotion Details
DWK Life Sciences
8264510004
4
Each for $161.29
Description
Description
  • All plugs are interchangeable with other HI-VAC counterparts
Specifications
Specifications
Glass
Replacement Plug
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'