DWK Life Sciences Kimble™ Wintrobe Erythrocyte Sedimentation Rate (ESR) Tubes
Offers full calibration
Supplier:  DWK Life Sciences 66F828SO
Catalog No. 02-681-1C
Explore available promotions
Description
Description
  • Cotton plugged
  • Full calibration in yellow
  • Fill with aspiration bulb No. 16
  • I.D. × L: 3 × 150mm (0.12 × 5.9 in.)
Specifications
Specifications
Soda Lime Glass
100/Pk.
15 cm
Wintrobe Pipet