missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ Clear Glass Straight-Sided Jars, Tall, Without Cap

Wide-mouth design

Supplier:  DWK Life Sciences 5510448B

 View more versions of this product

Catalog No. 13-774-84

Explore available promotions

Description

Description

  • Clear glass for easy viewing of contents
  • Type III soda-lime glass
  • Caps sold separately

All 32-ounce jars in this product family are packed in corrugated cartons with divider cells.

Specifications

Specifications

4 oz., 125 mL
4 oz.
2 in.
Cap
Bulk
Manufactured from Soda Lime Glass Conforming to USP Type III and ASTM E438, Type II Requirements
Round
Straight-sided Jar
48-400 mm
Cap
125 mL
Clear
Storage
Soda Lime Glass
Clear
Wide
102 mm
4.01 in.
51 mm
Videos
SDS
Documents

Documents

Promotions

Promotions