DWK Life Sciences Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes: 12.5mL Capacity
Tubes are made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class A or B glass requirements.
Supplier:  DWK Life Sciences 45170125
Catalog No. 05-603
Explore available promotions
Description
Description
Tubes are made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class A or B glass requirements.
  • Disposable
  • With conical shaped bottom
Specifications
Specifications
To contain
12.5 mL
Borosilicate Glass
0 to 1mL in 0.1%, >1 to 2mL in 0.15%, >2 to 5mL in 0.2%, >5 to 10mL in 0.4%, >10 to 25mL in 0.5%, >25mL in 1%
118 mm
Marking Spot
0 to 3 mL in 0.2 mL increments, 3 to 10 mL in 0.5 mL increments, 10 to 50 mL in 1 mL increments, 100 mL
2980 x g
USP, ASTM E 438, API 2542
Reusable
Used to determine the bottom sediment and water in petroleum
Conical
Conical
17 mm
Centrifuge Tube