DWK Life Sciences Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes: 13mL Capacity
Made with heavier walled tubing to withstand higher centrifugation speeds. Top is beaded for strength.
Supplier:  DWK Life Sciences 4517413
 View more versions of this product
Catalog No. 05-538-29A
Explore available promotions
Description
Description
  • Supplied with size 13 standard taper color-coded flathead PTFE stopper
  • Tube is plain, without a capacity scale
  • Do not centrifuge with stopper in tube
Specifications
Specifications
None
PTFE
Reusable
13
141 mm
17 mm
Centrifuge Tube
USP, ASTM E 438
13 mL
Borosilicate Glass
Conical
Conical
No
2050 x g