DWK Life Sciences KIMBLE™ EPA Water Analysis Vial, Without Closure
These clear and amber screw thread EPA vials for the collection and storage of water samples comply with the guidelines for establishing test procedures for the analysis of pollutants.
Supplier:  DWK Life Sciences 60958A16
 View more versions of this product
Catalog No. 03-339-5E
Explore available promotions
Description
Description
  • Clear EPA vials are Converted from Type I, 33 expansion borosilicate glass tubing and amber EPA vials are converted from Type I, 51 expansion borosilicate glass tubing
  • While vials are not pre-cleaned, they are packaged in a plastic shrink wrap module, lehr-clean, as they come from the end of the production lehr and then are placed into a corrugated shelf-pack
  • Open-top closures and PTFE-faced silicone septa or solid-top PFTE-faced white rubber-lined closures are available separately
Specifications
Specifications
1.18 in.
30 mm
60 mL
4.84 in.
2.02 oz.
123 mm
Clear
Borosilicate Glass
For water analysis
144
Screw Cap
EPA 601, 602, 603, 604
SDS
Documents
Documents
Accessories