DWK Life Sciences Kimble™ KG-35 Borosilicate Glass Serum Bottle, Aluminum Seal without Closure
Autoclavable Kimble™ serum bottles and vials are well-suited for the handling, containment and storage of a variety of liquids including reagents, vaccines, blood plasma, culture media, chromatography samples and more.
Supplier: DWK Life Sciences 61000G20
Description
- Ideal for packaging and storage where applications of injectable and parenteral solutions require utmost purity
- Highly resistant to thermal and mechanical shock as well as chemical attack
- Manufactured from USP Type I borosilicate molded glass
Specifications
0.67 oz., 20mL | |
0.67 oz. | |
Borosilicate Glass | |
Serum Bottle | |
33 mm |
20 mL | |
1.29 in. | |
59mm | |
2.32 in. |