DWK Life Sciences Kimble™ KIMAX™ Class A Mixing Bulb-Style Volumetric Flask with Standard Taper Stopper
Mixing bulb in neck contains approximately 0.1 of capacity of body to aid in dissolving or mixing contents
$1166.59 - $1166.59
Specifications
Calibration Calibrated To Contain
Closure Material Glass
Material Borosilicate Glass
Neck Style Bulb
Graduated Yes
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
K28019-100
View Documents Promotion Details
DWK Life Sciences
28019100
100 mL
Case of 4 Each for $1,166.59
Description
Description
Mixing bulb in neck contains approximately 0.1 of capacity of body to aid in dissolving or mixing contents
  • Calibrated “To Contain”
  • Designed from ASTM™ E 288 Class A unserialized requirements
  • With marking spot and standard taper 13 ground glass stopper
  • Tolerance ±0.08mL
Specifications
Specifications
Calibrated To Contain
Borosilicate Glass
Yes
Glass
Bulb
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'