missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ KIMAX™ Heavy-Duty Clear Glass Class A Volumetric Flasks with Glass Stoppers

Large numbers make it easy to identify volume from far away

Supplier:  DWK Life Sciences 92812G5

 View more versions of this product

Catalog No. 13-756-684


Explore available promotions

Description

Description

Large numbers make it easy to identify volume from far away

  • Wide Mouth
  • Heavy, uniform walls provide superior strength, durability and safety
  • Large, permanent, easy-to-read markings
  • Flat interior bottoms for stirring with standard stir bars
  • Graduated to Class A volumetric tolerances
  • Calibrated “To Contain”
  • Capacity:10 to 250mL
  • Calibrated to meet ASTM™ E288

Specifications

Specifications

Glass
Borosilicate Glass
9.7 cm
Yes
13
5 mL
3.8 in.
±0.08 mL
Wide
Videos
SDS
Documents

Documents

Promotions

Promotions