DWK Life Sciences Kimble™ Kontes™ Allihn Condensers with Straight Outlet Tube
For refluxing applications including Soxhlet extractions
$586.03 - $862.29
Specifications
Certifications/Compliance USP Type I and ASTM E438 Type I, Class A
Material Borosilicate Glass
Type Allihn Condenser
For Use With (Application) Refluxing operations, including soxhlet extractions
Includes Straight outlet tube, water cooled standard taper inner drip joint at bottom
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (English) Inner Tubing Standard Taper Joint No Height (English) Length (English) Jacket Price Quantity  
Catalog Number Mfr. No. Diameter (English) Inner Tubing Standard Taper Joint No Height (English) Length (English) Jacket Price Quantity  
K456000-0024
View Documents
DWK Life Sciences
4560000024
0.5 in. 71/60 16.9 in. 13.7 in.
Each for $862.29
K456000-0021
View Documents Promotion Details
DWK Life Sciences
4560000021
0.37 in. 34/45 10.8 in. 7.4 in.
Each for $586.03
K456000-0022
View Documents Promotion Details
DWK Life Sciences
4560000022
0.37 in. 45/50 14.3 in. 10.2 in.
Each for $619.41
Description
Description
  • Straight outlet tube at top Water-cooled standard taper inner drip joint at bottom
  • Accept No. 2 hose connections, except for 350mm size, which accepts No. 3
Specifications
Specifications
USP Type I and ASTM E438 Type I, Class A
Allihn Condenser
Straight outlet tube, water cooled standard taper inner drip joint at bottom
Borosilicate Glass
Refluxing operations, including soxhlet extractions
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'