DWK Life Sciences Kimble™ Kontes™ Friedrich Condensers
High-quality, water-cooled condensers to meet your vapor- or air-cooling needs
Supplier:  DWK Life Sciences 4370002440
 View more versions of this product
Catalog No. K437000-2440
Explore available promotions
Description
Description
Condensers provide three joint sizes to tackle a wide range of tasks.
  • Molded inner cold finger
  • Hose connection for 3/8 in. ID flexible tubing
24/40 Joint
  • Specially designed to provide a long vapor path, good heat transfer and anti-flooding characteristics
  • Inclined outer joint on the side and two hose connectors at the top
  • Inner drip joint at the bottom
  • Side arm is an outer standard taper joint and the bottom is an inner standard taper joint
45/50 Joint and 55/50 Joint
  • Used primarily in reflux mode and Soxhlet extractions
  • Water inlet and outlet located ring-sealed inner condensing tube
  • Inner drip joint and side outlet for drying tube
Specifications
Specifications
8.8 in.
Provide a long vapor path, good heat transfer and anti-flooding characteristics
Friedrich Condenser
Each
22.5 cm
24/40
325 mm
12.7 in.
Borosilicate Glass