Learn More
DWK Life Sciences Kimble™ Phenolic Caps with 14B White Rubber Liners
Resilient and resistant to moisture vapor; provides a chemical barrier
Supplier:  DWK Life Sciences 75204G89400
 View more versions of this product
Catalog No. 13-757-168
Only null left
Explore available promotions
Description
Description
  • Liner material: homogenous white rubber (0.050-inch)
  • Use for hermetic sealing for biologicals and other contents requiring sterilization
  • Autoclavable
Specifications
Specifications
Black
144 Cs.
Phenolic Cap
PTFE/14B Rubber
Phenolic Resin
89-400
Bottle
Product Suggestions
Product Suggestions