DWK Life Sciences Kimble™ Phenolic Caps with Solid PE Liners
Has good resistance to acids, alcohols, alkalis, aqueous solutions, oils, solvents
Supplier:  DWK Life Sciences 75202G22400
 View more versions of this product
Catalog No. 13-757-128
Explore available promotions
Alert:
Not recommended for hydrocarbon solvents.
Description
Description
  • General purpose phenolic screw cap with a solid polyethylene (PE) liner
  • Color: black
  • Liner material: solid polyethylene (0.040-in.)
  • Ideal for use with strong acid or base samples
  • Not recommended for hydrocarbon solvents
Specifications
Specifications
Black
144 Cs.
Phenolic Cap
Polyethylene
Phenolic Resin
22-400
Bottle