DWK Life Sciences Kimble™ Phenolic Caps with Solid PE Liners
Has good resistance to acids, alcohols, alkalis, aqueous solutions, oils, solvents
Supplier: DWK Life Sciences 75202G24400
Alert:
Not recommended for hydrocarbon solvents.
Description
- General purpose phenolic screw cap with a solid polyethylene (PE) liner
- Color: black
- Liner material: solid polyethylene (0.040-in.)
- Ideal for use with strong acid or base samples
- Not recommended for hydrocarbon solvents
Specifications
Black | |
144 Cs. | |
Phenolic Cap | |
Polyethylene |
Phenolic Resin | |
24-400 | |
Bottle |