DWK Life Sciences KIMBLE™ RAY-SORB™ Amber Erlenmeyer Flask
Protect solutions sensitive to light of the shorter wavelengths
$232.03 - $301.06
Specifications
Closure Material PTFE
Material Borosilicate Glass
Writing Surface Matte Finish Ceramic Enamel Marking
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
10-310-86
View Documents Promotion Details
DWK Life Sciences
2661050
50 mL
Case for $232.03
10-310-84
View Documents Promotion Details
DWK Life Sciences
26610125
125 mL
Case for $293.65
10-310-85
View Documents Promotion Details
DWK Life Sciences
26610250
250 mL
Case for $301.06
Description
Description
  • Features color-coded PTFE stopper
  • Capacity scale not present
Certifications
ASTM Specification E 1404, Type II, Class 2 requirements
Specifications
Specifications
PTFE
Borosilicate Glass
Matte Finish Ceramic Enamel Marking
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'