DWK Life Sciences Kimble™ Plastic Bumper for 100mL Cylinders
When placed around a cylinder near the top, this sturdy split ring protects against breakage if the cylinder is upset.
Description
- 28.5-31.0mm (100mL)
- Made of yellow low density polyethylene
- Position of ring can be adjusted to avoid contact with the top of a receiver when liquid is poured from the cylinder
- Should a ring become loose after prolonged use, soak it briefly in hot water (90°C) to return it to its original shape
Specifications
Polyethylene | |
Plastic Bumper | |
Bumper |