DWK Life Sciences Kimble™ Screw Caps for Disposable Glass Tubes
Linerless, one-piece closures with inner sealing ring
$198.09 - $309.74
Specifications
Product Type | Screw Cap |
---|---|
Certifications/Compliance | ASTM E982 |
Autoclavable | Autoclavable |
Material | Polypropylene |
Catalog Number | Mfr. No. | Closure Size | Color | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Closure Size | Color | Price | Quantity | ||||
03-340-77C
|
DWK Life Sciences
73805B13415 |
13-415 | Natural |
Case of 1000 Each for $291.38
|
|
||||
03-340-77E
|
DWK Life Sciences
73805B18415 |
18-415 | Natural |
Case of 500 Each for $198.09
|
|
||||
03-340-77D
|
DWK Life Sciences
73805B15415 |
15-415 | White |
Case of 1000 Each for $309.74
|
|
||||
Description
Manufactured from polypropylene. Kimble Chase™ Screw Caps for Disposable Glass Tubes are autoclavable.
- Polypropylene
- Autoclavable
Compliance
Designed from ASTM™ E 982
Specifications
Screw Cap | |
ASTM E982 | |
Autoclavable | |
Polypropylene |