DWK Life Sciences Kimble™ Solid Borosilicate Glass Beads for Distillation Columns
Resist attack by liquids or vapors
Supplier:  DWK Life Sciences 135003
 View more versions of this product
Catalog No. 10-310-1
Explore available promotions
Description
Description
May be used as mixing beads, boiling stones, or packing
Order Info
Packed in 1 lb. boxes
Specifications
Specifications
Borosilicate Glass
3 mm