Ansell Kimtech™ A7 Certified Liquid Barrier Gowns
Non-sterile gowns deliver stringent barrier protection and provide contamination control for clinical, chemotherapy, and laboratory settings.
Supplier:  Ansell 47994
 View more versions of this product
Catalog No. 17-986-052
Explore available promotions
Description
Description
  • Liquid barrier gowns
  • Seamless front design
  • Easy to don-and-doff design to reduce contamination risk
  • Material offers enhanced liquid barrier protection
  • Fabric tested for blood and blood-borne pathogen penetration, chemotherapy drug and hormone permeation
  • Passes ASTM F1670 and ASTM F1671
  • Built-in thumb loops help maintain glove and garment interface
  • Low lint to minimize contamination
  • Meets USP 800 guidelines
  • Bulk-packaged and available in a range of sizes
Compliance
Meets USP 800 guidelines
Specifications
Specifications
Controlled Environments Gown
None
10 Bags of 10 Gowns
Blue
Thumb Loop
Seamless Front
Not Autoclavable
Clinical, Pharmaceutical, Laboratories, Chemotherapy
Unisex
Polyethylene, Polypropylene
Single-use
47994
ASTM 1670, ASTM 1671, Meets USP 800 Guidelines
Product Suggestions
Product Suggestions